Product Details

SNP ID
rs480498
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:179883961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGTTTGTGGTAGACTATTGATA[C/G]GCGGAGGAGTGGGTTTTCAGCTTAG
Phenotype
MIM: 614512 MIM: 614513
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TOR1AIP1 PubMed Links

Gene Details

Gene
TOR1AIP1
Gene Name
torsin 1A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267578.1 Intron NP_001254507.1
NM_015602.3 Intron NP_056417.2
XM_011509403.1 Intron XP_011507705.1
XM_011509404.1 Intron XP_011507706.1
Gene
TOR1AIP2
Gene Name
torsin 1A interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details