Product Details
- SNP ID
-
rs362108
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:18118036 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTGCGTCCCATCAGGAGGTCCTC[A/G]ATGTCGGGTTGGCCCACAGTTAGCG
- Phenotype
-
MIM: 605742
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA8
PubMed Links
Gene Details
- Gene
- TUBA8
- Gene Name
- tubulin alpha 8
View Full Product Details