Product Details

SNP ID
rs243337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4402623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAATTGGCGCGCGGACTGCGGGG[A/G]TCGCCGCGGTTCCCGCCAAATACGA
Phenotype
MIM: 601246 MIM: 601768
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHAF1A PubMed Links

Gene Details

Gene
CHAF1A
Gene Name
chromatin assembly factor 1 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005483.2 28 Intron NP_005474.2
XM_011527605.2 28 UTR 5 XP_011525907.1
XM_011527606.2 28 UTR 5 XP_011525908.1
XM_011527607.2 28 UTR 5 XP_011525909.1
Gene
SH3GL1
Gene Name
SH3 domain containing GRB2 like 1, endophilin A2
There are no transcripts associated with this gene.

View Full Product Details