Product Details

SNP ID
rs225626
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:142151157 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTCATCTTGGATTTTATCAAGAGG[G/T]TGAGGAGGAACAGTTTGGGGCCAAG
Phenotype
MIM: 162341 MIM: 610902
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NMBR PubMed Links

Gene Details

Gene
NMBR
Gene Name
neuromedin B receptor
There are no transcripts associated with this gene.

Gene
VTA1
Gene Name
vesicle trafficking 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286371.1 Intron NP_001273300.1
NM_001286372.1 Intron NP_001273301.1
NM_016485.4 Intron NP_057569.2

View Full Product Details