Product Details

SNP ID
rs292556
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:135186671 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATCTGCTTATTTGTTGAAGGTTCA[C/T]GTCCCATACTGGGGAATGCAAGCTG
Phenotype
MIM: 616303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR91 PubMed Links

Gene Details

Gene
WDR91
Gene Name
WD repeat domain 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014149.3 Intron NP_054868.3
XM_011516083.2 Intron XP_011514385.1
XM_011516086.2 Intron XP_011514388.1
XM_017012070.1 Intron XP_016867559.1
XM_017012071.1 Intron XP_016867560.1
XM_017012072.1 Intron XP_016867561.1

View Full Product Details