Product Details

SNP ID
rs743273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24116629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTTAGTAGGTCATGTCCACAGC[A/G]TATTGGTCTTGCTCTTGTTCTAAGG
Phenotype
MIM: 613317 MIM: 162080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF11 PubMed Links

Gene Details

Gene
DCAF11
Gene Name
DDB1 and CUL4 associated factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163484.1 Intron NP_001156956.1
NM_025230.4 Intron NP_079506.3
NM_181357.2 Intron NP_852002.1
Gene
NRL
Gene Name
neural retina leucine zipper
There are no transcripts associated with this gene.

View Full Product Details