Product Details
- SNP ID
-
hCV2410844
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:111780229 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCAAGTCACAATAGGAATATTC[A/C]ATAAAGGTACCCTTTCTCTCTTTTT
- Phenotype
-
MIM: 606941
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ALG9
PubMed Links
Gene Details
- Gene
- ALG9
- Gene Name
- ALG9, alpha-1,2-mannosyltransferase
There are no transcripts associated with this gene.
View Full Product Details