Product Details

SNP ID
rs597306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219629396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGAACCTGTGGAGCCCCCCC[A/C]CTCAGGGACCCCACAGAAGGCAAAG
Phenotype
MIM: 106195
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC4A3 PubMed Links

Gene Details

Gene
SLC4A3
Gene Name
solute carrier family 4 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326559.1 870 Missense Mutation CAC,CCC H157P NP_001313488.1
NM_005070.3 870 Missense Mutation CAC,CCC H157P NP_005061.2
NM_201574.2 870 Missense Mutation CAC,CCC H157P NP_963868.2
XM_005246789.4 870 Missense Mutation CAC,CCC H157P XP_005246846.1
XM_005246790.4 870 Intron XP_005246847.1
XM_011511665.2 870 Missense Mutation CAC,CCC H104P XP_011509967.1
XM_011511667.2 870 Intron XP_011509969.1

View Full Product Details