Product Details
- SNP ID
-
rs16906093
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:90025874 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCTGACGAATATTCAGAAGTGCT[A/T]CCTTTGCTCTTTTGAGTAACTAAAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4290
PubMed Links
Gene Details
- Gene
- MIR4290
- Gene Name
- microRNA 4290
There are no transcripts associated with this gene.
- Gene
- MIR4290HG
- Gene Name
- MIR4290 host gene
There are no transcripts associated with this gene.
View Full Product Details