Product Details
- SNP ID
-
rs2836161
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:38124267 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTGATTAGAGCAATGAGTTCCAACT[C/T]GGCAGTTTTTGGAAACTGAACATGA
- Phenotype
-
MIM: 604829
MIM: 613396
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DSCR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60876887] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DSCR4
- Gene Name
- Down syndrome critical region 4
There are no transcripts associated with this gene.
- Gene
- DSCR8
- Gene Name
- Down syndrome critical region 8
There are no transcripts associated with this gene.
View Full Product Details