Product Details

SNP ID
rs7283354
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:33504763 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGCTGGGGTGCTCTGAACAGACA[C/T]TCATGGTGACAGGAAGGTGGGGAGT
Phenotype
MIM: 138440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GART PubMed Links

Gene Details

Gene
GART
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000819.4 Intron NP_000810.1
NM_001136005.1 Intron NP_001129477.1
NM_001136006.1 Intron NP_001129478.1
NM_175085.2 Intron NP_780294.1
XM_005260941.1 Intron XP_005260998.1
XM_006723989.1 Intron XP_006724052.1
XM_006723990.1 Intron XP_006724053.1
XM_011529526.1 Intron XP_011527828.1

View Full Product Details