Product Details

SNP ID
rs12483711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34731139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTGTTTGACAATATGAAACATA[C/T]GGATTTCAGATCCCCTCTTCATAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00160 PubMed Links

Gene Details

Gene
LINC00160
Gene Name
long intergenic non-protein coding RNA 160
There are no transcripts associated with this gene.

View Full Product Details