Product Details

SNP ID
rs8131610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25650421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCCAAGAATCTCATTCAGCAT[C/T]AGGCAAATGCCAAAATCAGATACAT
Phenotype
MIM: 606870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JAM2 PubMed Links

Gene Details

Gene
JAM2
Gene Name
junctional adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270407.1 Intron NP_001257336.1
NM_001270408.1 Intron NP_001257337.1
NM_021219.3 Intron NP_067042.1

View Full Product Details