Product Details
- SNP ID
-
rs9267525
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31640165 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCAGGCATGACGGGGAAACCTGGA[C/T]AGAGAGAGAGGCTTAGGGAAGAGGA
- Phenotype
-
MIM: 142590
MIM: 142580
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BAG6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202172137] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAG6
- Gene Name
- BCL2 associated athanogene 6
- Gene
- MIR6832
- Gene Name
- microRNA 6832
There are no transcripts associated with this gene.
- Gene
- PRRC2A
- Gene Name
- proline rich coiled-coil 2A
There are no transcripts associated with this gene.
View Full Product Details