Product Details

SNP ID
rs2269711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31173835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCCACCTCTCCCCACCAGGTG[A/T]GGCGGCTGGCATGGCTGGGTGGGGA
Phenotype
MIM: 164177 MIM: 600912
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
POU5F1 PubMed Links

Gene Details

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
There are no transcripts associated with this gene.

Gene
PSORS1C3
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TCF19
Gene Name
transcription factor 19
There are no transcripts associated with this gene.

View Full Product Details