Product Details

SNP ID
rs11089791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36470273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAGCAGGTCACAGGACACAGTC[C/T]TTGTCCTTGTTAGTGCCTCAGGCCC
Phenotype
MIM: 609063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXN2 PubMed Links

Gene Details

Gene
TXN2
Gene Name
thioredoxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012473.3 Intron NP_036605.2
XM_005261508.1 Intron XP_005261565.1
XM_006724226.1 Intron XP_006724289.1
XM_011530111.2 Intron XP_011528413.1

View Full Product Details