Product Details

SNP ID
rs1207492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47837154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCATTCTAAGCCCCATTGGGTTTG[A/G]AGCATTATGTTTTTTAAGAGTGTTA
Phenotype
MIM: 314998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF81 PubMed Links

Gene Details

Gene
ZNF81
Gene Name
zinc finger protein 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007137.3 Intron NP_009068.2
XM_005272600.3 Intron XP_005272657.1
XM_011543899.2 Intron XP_011542201.1
XM_011543900.2 Intron XP_011542202.1
XM_017029486.1 Intron XP_016884975.1
XM_017029487.1 Intron XP_016884976.1

View Full Product Details