Product Details

SNP ID
rs11091212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47844711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGATATCTATGAGTGGAATTTCT[G/T]GGTTATATGGTGACTTTATGGTTGA
Phenotype
MIM: 314998
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF81 PubMed Links

Gene Details

Gene
ZNF81
Gene Name
zinc finger protein 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007137.3 910 Intron NP_009068.2
XM_005272600.3 910 Intron XP_005272657.1
XM_011543899.2 910 UTR 5 XP_011542201.1
XM_011543900.2 910 Intron XP_011542202.1
XM_017029486.1 910 Intron XP_016884975.1
XM_017029487.1 910 Intron XP_016884976.1

View Full Product Details