Product Details

SNP ID
rs34834819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37342353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATTGCCCTTTCTCTAGCCTCC[C/T]TCTAGCAGCACCTGTTCCCCAGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELFN2 PubMed Links

Gene Details

Gene
ELFN2
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052906.4 Intron NP_443138.2
Gene
LOC100506271
Gene Name
uncharacterized LOC100506271
There are no transcripts associated with this gene.

View Full Product Details