Product Details

SNP ID
rs6953256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:91946103 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTACAAAAAAAAGGATGGTGCT[C/T]CATGCAAGCTGTATATTGATTATTG
Phenotype
MIM: 604001
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP9 PubMed Links

Gene Details

Gene
AKAP9
Gene Name
A-kinase anchoring protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005751.4 Intron NP_005742.4
NM_147185.2 Intron NP_671714.1
XM_017011642.1 Intron XP_016867131.1
XM_017011643.1 Intron XP_016867132.1
XM_017011644.1 Intron XP_016867133.1
XM_017011645.1 Intron XP_016867134.1
XM_017011646.1 Intron XP_016867135.1
XM_017011647.1 Intron XP_016867136.1
XM_017011648.1 Intron XP_016867137.1
XM_017011649.1 Intron XP_016867138.1
XM_017011650.1 Intron XP_016867139.1
XM_017011651.1 Intron XP_016867140.1
XM_017011652.1 Intron XP_016867141.1
XM_017011653.1 Intron XP_016867142.1
XM_017011654.1 Intron XP_016867143.1
XM_017011655.1 Intron XP_016867144.1
XM_017011656.1 Intron XP_016867145.1
XM_017011657.1 Intron XP_016867146.1
XM_017011658.1 Intron XP_016867147.1
XM_017011659.1 Intron XP_016867148.1
XM_017011660.1 Intron XP_016867149.1

View Full Product Details