Product Details

SNP ID
rs9276430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32744405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGTTGAATCAGAACACTATAGA[C/T]ATTTAGTAACCTCCTTCAGAGGAAA
Phenotype
MIM: 613503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DQA2 PubMed Links

Gene Details

Gene
HLA-DQA2
Gene Name
major histocompatibility complex, class II, DQ alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020056.4 Intron NP_064440.1
Gene
MIR3135B
Gene Name
microRNA 3135b
There are no transcripts associated with this gene.

View Full Product Details