Product Details

SNP ID
rs10438012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23322013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGCTTGATTCGGGCGTCACGG[G/T]TGCCTAGTGTTGTTCTAGAGAGGGT
Phenotype
MIM: 602279
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL2L2-PABPN1 PubMed Links

Gene Details

Gene
BCL2L2-PABPN1
Gene Name
BCL2L2-PABPN1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199864.1 Intron NP_001186793.1
Gene
PABPN1
Gene Name
poly(A) binding protein nuclear 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004643.3 Intron NP_004634.1

View Full Product Details