Product Details

SNP ID
rs6010202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50205748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACATGAGGCCGAGGCCGGGGACCC[A/G]AAGACCTGTGGGGAAGGGAATGCGA
Phenotype
MIM: 607917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELO PubMed Links

Gene Details

Gene
SELO
Gene Name
selenoprotein O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031454.1 Intron NP_113642.1
Gene
TRABD
Gene Name
TraB domain containing
There are no transcripts associated with this gene.

View Full Product Details