Product Details
- SNP ID
-
rs2142551
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:45286394 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCCATTACAGAGCAGGGAAGCAAG[C/G]CTCAGAGAGGTGGAATAACCCCCCC
- Phenotype
-
MIM: 611559
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UPK3A
PubMed Links
Gene Details
- Gene
- UPK3A
- Gene Name
- uroplakin 3A
View Full Product Details