Product Details
- SNP ID
-
rs760951
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
63
- Location
-
Chr.1:209688180 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGCTCTAGCCATTCAGGCTACCT[A/G]AGTTGCCTCCAAAATGCCATGCTTT
- Phenotype
-
MIM: 600713
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD11B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75744075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD11B1
- Gene Name
- hydroxysteroid 11-beta dehydrogenase 1
- Gene
- LOC101930114
- Gene Name
- uncharacterized LOC101930114
There are no transcripts associated with this gene.
View Full Product Details