Product Details

SNP ID
rs2903781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45307570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCACCTCCCATACTTTCCTACC[C/T]GGAGATGCCTCTGGGCCCCTAAATC
Phenotype
MIM: 603897 MIM: 616104
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MATN4 PubMed Links

Gene Details

Gene
MATN4
Gene Name
matrilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003833.4 Intron NP_003824.2
NM_030590.3 Intron NP_085080.1
NM_030592.3 Intron NP_085095.1
XM_005260597.1 Intron XP_005260654.1
XM_017028113.1 Intron XP_016883602.1
XM_017028114.1 Intron XP_016883603.1
XM_017028115.1 Intron XP_016883604.1
Gene
RBPJL
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281448.1 Intron NP_001268377.1
NM_001281449.1 Intron NP_001268378.1
NM_014276.3 Intron NP_055091.2
XM_011528521.2 Intron XP_011526823.1
XM_011528522.2 Intron XP_011526824.1
XM_011528523.2 Intron XP_011526825.1

View Full Product Details