Product Details

SNP ID
rs3789334
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:6772275 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTATAAAATCAGAATTTATATCTTT[A/G]TGAGTGTAAATGTTACCTGAGAAAC
Phenotype
MIM: 112261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP2 PubMed Links

Gene Details

Gene
BMP2
Gene Name
bone morphogenetic protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200.3 Intron NP_001191.1
XM_011529323.2 Intron XP_011527625.1

View Full Product Details