Product Details

SNP ID
rs2267424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41093522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTTTCCTCTACTTTCCACTCTT[A/C]GAAATTTTCTCTCCTTTGCAGAATA
Phenotype
MIM: 602700
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EP300 PubMed Links

Gene Details

Gene
EP300
Gene Name
E1A binding protein p300
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001429.3 Intron NP_001420.2
XM_006724165.3 Intron XP_006724228.1
Gene
MIR1281
Gene Name
microRNA 1281
There are no transcripts associated with this gene.

View Full Product Details