Product Details

SNP ID
rs6297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:77462224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCGCTTAGGCGACCCCACTGCAAA[C/T]GGCAAGTCAACTTGTGCACTTAAAA
Phenotype
MIM: 182131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR1B PubMed Links

Gene Details

Gene
HTR1B
Gene Name
5-hydroxytryptamine receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000863.2 1799 UTR 3 NP_000854.1

View Full Product Details