Product Details

SNP ID
rs11905265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13109252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACAAGATTATTCTGCCAGACATT[C/T]ATCCTCCCTTGTTCTACATCACCAT
Phenotype
MIM: 611120 MIM: 608270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPTLC3 PubMed Links

Gene Details

Gene
SPTLC3
Gene Name
serine palmitoyltransferase long chain base subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018327.2 Intron NP_060797.2
XM_011529279.1 Intron XP_011527581.1
XM_011529280.2 Intron XP_011527582.1
XM_017027922.1 Intron XP_016883411.1
Gene
TASP1
Gene Name
taspase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323602.1 Intron NP_001310531.1
NM_001323603.1 Intron NP_001310532.1
NM_001323604.1 Intron NP_001310533.1
NM_017714.2 Intron NP_060184.2
XM_011529292.1 Intron XP_011527594.1
XM_011529294.1 Intron XP_011527596.1
XM_017027927.1 Intron XP_016883416.1
XM_017027928.1 Intron XP_016883417.1
XM_017027929.1 Intron XP_016883418.1
XM_017027930.1 Intron XP_016883419.1
XM_017027931.1 Intron XP_016883420.1

View Full Product Details