Product Details

SNP ID
rs34615149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2925758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCCGGGGCTGGGGGAGGTGT[A/G]GTGGGACTGGCCCTCCTGGTACCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLYWCH1 PubMed Links

Gene Details

Gene
FLYWCH1
Gene Name
FLYWCH-type zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308068.1 Intron NP_001294997.1
NM_020912.1 Intron NP_065963.1
NM_032296.2 Intron NP_115672.2
XM_006720962.3 Intron XP_006721025.1
XM_011522701.2 Intron XP_011521003.1
XM_011522702.2 Intron XP_011521004.1
XM_011522703.2 Intron XP_011521005.1
XM_011522704.2 Intron XP_011521006.1
XM_011522705.2 Intron XP_011521007.1
XM_011522707.2 Intron XP_011521009.1
XM_017023772.1 Intron XP_016879261.1

View Full Product Details