Product Details

SNP ID
rs2645482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59865754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACTGAACTTGCAGTGAGGTGTG[G/T]CACAGAACTCAGTACCAGTCAATGG
Phenotype
MIM: 607344
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TUBD1 PubMed Links

Gene Details

Gene
TUBD1
Gene Name
tubulin delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193609.1 Intron NP_001180538.1
NM_001193610.1 Intron NP_001180539.1
NM_001193611.1 Intron NP_001180540.1
NM_001193612.1 Intron NP_001180541.1
NM_001193613.1 Intron NP_001180542.1
NM_016261.3 Intron NP_057345.2
XM_005257425.1 Intron XP_005257482.1
XM_011524871.2 Intron XP_011523173.1
XM_017024716.1 Intron XP_016880205.1
XM_017024717.1 Intron XP_016880206.1
XM_017024718.1 Intron XP_016880207.1
XM_017024719.1 Intron XP_016880208.1
XM_017024720.1 Intron XP_016880209.1

View Full Product Details