Product Details

SNP ID
rs11823045
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:17552020 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAAGCAGGCTGAAGCCCCAGACTC[C/T]GTGGCCATGTCTTCCTGGGAAAGGC
Phenotype
MIM: 604487 MIM: 605242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTOG PubMed Links

Gene Details

Gene
OTOG
Gene Name
otogelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277269.1 273 Silent Mutation TCC,TCT S91S NP_001264198.1
NM_001292063.1 273 Silent Mutation TCC,TCT S79S NP_001278992.1
Gene
USH1C
Gene Name
USH1 protein network component harmonin
There are no transcripts associated with this gene.

View Full Product Details