Product Details
- SNP ID
-
rs12451097
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:4959667 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACTAGGGGCAGAGAGGAGGGTAGG[A/G]CGGGCCTAGAAATCCGTACCTCAGC
- Phenotype
-
MIM: 611508
MIM: 131370
MIM: 610056
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMTA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80259031] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMTA2
- Gene Name
- calmodulin binding transcription activator 2
There are no transcripts associated with this gene.
- Gene
- ENO3
- Gene Name
- enolase 3
There are no transcripts associated with this gene.
- Gene
- SPAG7
- Gene Name
- sperm associated antigen 7
View Full Product Details