Product Details

SNP ID
rs950540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89211633 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGGCTGTCACTGCTCTTTATGG[C/T]GCGAGGTGGTCCAACTTCTCAGTTC
Phenotype
MIM: 612196 MIM: 180090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD2 PubMed Links

Gene Details

Gene
ABHD2
Gene Name
abhydrolase domain containing 2
There are no transcripts associated with this gene.

Gene
RLBP1
Gene Name
retinaldehyde binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000326.4 Intron NP_000317.1
XM_011521870.2 Intron XP_011520172.1
XM_017022460.1 Intron XP_016877949.1

View Full Product Details