Product Details

SNP ID
rs6022866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:53946296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCACCGCAGACCCTGGCAAGCAC[C/G]GATCTGTTTTCATCCCTAGAGTTCT
Phenotype
MIM: 602968
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCAS1 PubMed Links

Gene Details

Gene
BCAS1
Gene Name
breast carcinoma amplified sequence 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316361.2 Intron NP_001303290.1
NM_001323347.1 Intron NP_001310276.1
NM_003657.3 Intron NP_003648.2
XM_005260587.2 Intron XP_005260644.1
XM_005260589.2 Intron XP_005260646.1
XM_005260590.2 Intron XP_005260647.1
XM_005260591.2 Intron XP_005260648.1
XM_005260593.2 Intron XP_005260650.1
XM_005260594.2 Intron XP_005260651.1
XM_005260595.2 Intron XP_005260652.1
XM_011529090.2 Intron XP_011527392.1
XM_011529091.2 Intron XP_011527393.1
XM_017028110.1 Intron XP_016883599.1
XM_017028111.1 Intron XP_016883600.1
XM_017028112.1 Intron XP_016883601.1

View Full Product Details