Product Details

SNP ID
rs361640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20399325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATCATTATGTAATATTCTCTTT[A/G]TCTCTGATAGTCCTCTCTCTGAAAT
Phenotype
MIM: 194548
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP41 PubMed Links

Gene Details

Gene
USP41
Gene Name
ubiquitin specific peptidase 41
There are no transcripts associated with this gene.

Gene
ZNF74
Gene Name
zinc finger protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256523.1 Intron NP_001243452.1
NM_001256524.1 Intron NP_001243453.1
NM_001256525.1 Intron NP_001243454.1
NM_003426.3 Intron NP_003417.2

View Full Product Details