Product Details
- SNP ID
-
rs9900415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:2693533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGGGCACCCAGAACCCCACCAC[A/G]GCCTCCACCAGCCCCGGCCAGGCAG
- Phenotype
-
MIM: 616184
MIM: 601545
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLUH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs568499003] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLUH
- Gene Name
- clustered mitochondria homolog
- Gene
- MIR6776
- Gene Name
- microRNA 6776
There are no transcripts associated with this gene.
- Gene
- PAFAH1B1
- Gene Name
- platelet activating factor acetylhydrolase 1b regulatory subunit 1
There are no transcripts associated with this gene.
View Full Product Details