Product Details

SNP ID
rs11880094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39835000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAAGAGTTGAGAGGTGGGACCTT[C/T]GAGAGATGATTCAGGTCATGAGGCC
Phenotype
MIM: 604556 MIM: 134795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK1B PubMed Links

Gene Details

Gene
DYRK1B
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004714.2 Intron NP_004705.1
NM_006483.2 Intron NP_006474.1
NM_006484.2 Intron NP_006475.1
XM_005259398.4 Intron XP_005259455.1
Gene
FBL
Gene Name
fibrillarin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001436.3 Intron NP_001427.2
XM_005258651.2 Intron XP_005258708.1
XM_011526623.2 Intron XP_011524925.1
Gene
MIR6719
Gene Name
microRNA 6719
There are no transcripts associated with this gene.

View Full Product Details