Product Details

SNP ID
rs4925067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:20017909 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGATTCAGTGTCCCTGTTTCAT[C/G]ATACTGAATAAAAACTTGGTGATTC
Phenotype
MIM: 608793
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPECC1 PubMed Links

Gene Details

Gene
SPECC1
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033553.2 Intron NP_001028725.1
NM_001033554.2 Intron NP_001028726.1
NM_001033555.2 Intron NP_001028727.1
NM_001243438.1 Intron NP_001230367.1
NM_001243439.1 Intron NP_001230368.1
NM_152904.4 Intron NP_690868.3
XM_005256860.2 Intron XP_005256917.1
XM_011524075.1 Intron XP_011522377.1
XM_017025317.1 Intron XP_016880806.1
XM_017025318.1 Intron XP_016880807.1
XM_017025319.1 Intron XP_016880808.1
XM_017025320.1 Intron XP_016880809.1
XM_017025321.1 Intron XP_016880810.1
XM_017025322.1 Intron XP_016880811.1
XM_017025323.1 Intron XP_016880812.1
XM_017025324.1 Intron XP_016880813.1
XM_017025325.1 Intron XP_016880814.1

View Full Product Details