Product Details

SNP ID
rs917585
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:150193386 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGACAGCAAGATTATGACTTTCTGA[C/G]ACAGATCACGGCTTAAGTACCTTCT
Phenotype
MIM: 600746 MIM: 606205
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDX1 PubMed Links

Gene Details

Gene
CDX1
Gene Name
caudal type homeobox 1
There are no transcripts associated with this gene.

Gene
SLC6A7
Gene Name
solute carrier family 6 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014228.4 Intron NP_055043.2
XM_017009767.1 Intron XP_016865256.1
XM_017009768.1 Intron XP_016865257.1
XM_017009769.1 Intron XP_016865258.1
XM_017009770.1 Intron XP_016865259.1

View Full Product Details