Product Details

SNP ID
rs2525881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1998103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCTCCCTCAGGGTCCAGGAC[C/T]TCTCCCTCCCAGACCACTGTCTCCC
Phenotype
MIM: 103280 MIM: 615509 MIM: 600789
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H19 PubMed Links

Gene Details

Gene
H19
Gene Name
H19, imprinted maternally expressed transcript (non-protein coding)
There are no transcripts associated with this gene.

Gene
HOTS
Gene Name
H19 opposite tumor suppressor
There are no transcripts associated with this gene.

Gene
LINC01219
Gene Name
long intergenic non-protein coding RNA 1219
There are no transcripts associated with this gene.

Gene
MIR675
Gene Name
microRNA 675
There are no transcripts associated with this gene.

Gene
MRPL23
Gene Name
mitochondrial ribosomal protein L23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021134.3 Intron NP_066957.3
XM_006718271.2 Intron XP_006718334.1
XM_011520273.1 Intron XP_011518575.1
XM_011520275.2 Intron XP_011518577.1
XM_011520276.2 Intron XP_011518578.1
Gene
MRPL23-AS1
Gene Name
MRPL23 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details