Product Details

SNP ID
rs12980989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35613358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCTCTCCCGCGCTCCCCCACC[A/C]CCCGCCTTCCCAGGGAATGAGAATA
Phenotype
MIM: 613432
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAUS5 PubMed Links

Gene Details

Gene
HAUS5
Gene Name
HAUS augmin like complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015302.1 Intron NP_056117.1
XM_011526684.2 Intron XP_011524986.1

View Full Product Details