Product Details

SNP ID
rs2624841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50160982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTTTGGGCTGGAAAGCACCCC[C/T]CTTCCCCGGGCCCCAGGAAGGTGAG
Phenotype
MIM: 601124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107986035 PubMed Links

Gene Details

Gene
LOC107986035
Gene Name
basic proline-rich protein-like
There are no transcripts associated with this gene.

Gene
SEMA3F
Gene Name
semaphorin 3F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318798.1 Intron NP_001305727.1
NM_001318800.1 Intron NP_001305729.1
NM_004186.4 Intron NP_004177.3
XM_005265381.4 Intron XP_005265438.1
XM_005265382.4 Intron XP_005265439.1
XM_006713289.3 Intron XP_006713352.1
XM_006713290.3 Intron XP_006713353.1
XM_011533998.2 Intron XP_011532300.1
XM_011534000.2 Intron XP_011532302.1
Gene
SEMA3F-AS1
Gene Name
SEMA3F antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details