Product Details

SNP ID
rs5986916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153419527 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGCATGCGTGAAACAGAGTGGGAC[C/T]GGGCCTCTGGCTTTTCCCCCAGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP92 PubMed Links

Gene Details

Gene
ZFP92
Gene Name
ZFP92 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136273.1 Intron NP_001129745.1
XM_005274652.3 Intron XP_005274709.1
XM_011531115.2 Intron XP_011529417.1
XM_017029281.1 Intron XP_016884770.1

View Full Product Details