Product Details

SNP ID
rs11737447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7758249 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAACTCCTATTTAAACATGAGA[A/G]CCTAGTTCAAATAACCCCCACTCCT
Phenotype
MIM: 608252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFAP1 PubMed Links

Gene Details

Gene
AFAP1
Gene Name
actin filament associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134647.1 Intron NP_001128119.1
NM_198595.2 Intron NP_940997.1
XM_006713909.3 Intron XP_006713972.1
XM_011513544.2 Intron XP_011511846.1
XM_017008535.1 Intron XP_016864024.1
XM_017008536.1 Intron XP_016864025.1
Gene
AFAP1-AS1
Gene Name
AFAP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details