Product Details

SNP ID
rs4728467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139435816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAATCCATTGTAAGGGTGCCCC[A/G]CCGCTTACCCACTTAAAGTCATGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRG2 PubMed Links

Gene Details

Gene
KLRG2
Gene Name
killer cell lectin like receptor G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198508.2 Intron NP_940910.1
XM_005250311.3 Intron XP_005250368.1
XM_011516140.2 Intron XP_011514442.1
XM_011516141.2 Intron XP_011514443.1
Gene
LOC100129148
Gene Name
uncharacterized LOC100129148
There are no transcripts associated with this gene.

View Full Product Details