Product Details

SNP ID
rs12811969
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:1621902 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCACGAACAAGCAATTCTGAAAACA[G/T]TGGAAATTATTGAAGTCCTCCTTCA
Phenotype
MIM: 606361
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
WNT5B PubMed Links

Gene Details

Gene
WNT5B
Gene Name
Wnt family member 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030775.2 Intron NP_110402.2
NM_032642.2 Intron NP_116031.1

View Full Product Details