Product Details

SNP ID
rs304303
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:43712399 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTATGTGAGTGTATACATTTCT[G/T]TCTCTCCTACTAGACCAGGTGCTCT
Phenotype
MIM: 609764 MIM: 606494
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
There are no transcripts associated with this gene.

Gene
KDM4A-AS1
Gene Name
KDM4A antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101929592
Gene Name
uncharacterized LOC101929592
There are no transcripts associated with this gene.

Gene
ST3GAL3
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270459.1 Intron NP_001257388.1
NM_001270460.1 Intron NP_001257389.1
NM_001270461.1 Intron NP_001257390.1
NM_001270462.1 Intron NP_001257391.1
NM_001270463.1 Intron NP_001257392.1
NM_001270464.1 Intron NP_001257393.1
NM_001270465.1 Intron NP_001257394.1
NM_001270466.1 Intron NP_001257395.1
NM_006279.3 Intron NP_006270.1
NM_174963.3 Intron NP_777623.2
NM_174964.2 Intron NP_777624.1
NM_174965.2 Intron NP_777625.1
NM_174966.2 Intron NP_777626.1
NM_174967.2 Intron NP_777627.1
NM_174968.3 Intron NP_777628.2
NM_174969.2 Intron NP_777629.1
NM_174970.2 Intron NP_777630.1
NM_174971.3 Intron NP_777631.2
XM_006710825.3 Intron XP_006710888.1
XM_006710827.3 Intron XP_006710890.1
XM_006710828.3 Intron XP_006710891.1
XM_011541973.2 Intron XP_011540275.1
XM_011541974.2 Intron XP_011540276.1
XM_011541976.2 Intron XP_011540278.1
XM_011541977.2 Intron XP_011540279.1
XM_011541980.2 Intron XP_011540282.1
XM_011541981.2 Intron XP_011540283.1
XM_011541983.2 Intron XP_011540285.1
XM_011541984.2 Intron XP_011540286.1
XM_011541985.2 Intron XP_011540287.1
XM_011541986.2 Intron XP_011540288.1
XM_011541987.2 Intron XP_011540289.1
XM_011541988.2 Intron XP_011540290.1
XM_017002109.1 Intron XP_016857598.1
XM_017002110.1 Intron XP_016857599.1
XM_017002111.1 Intron XP_016857600.1
XM_017002112.1 Intron XP_016857601.1
XM_017002113.1 Intron XP_016857602.1
XM_017002114.1 Intron XP_016857603.1
XM_017002115.1 Intron XP_016857604.1
XM_017002116.1 Intron XP_016857605.1
XM_017002117.1 Intron XP_016857606.1
XM_017002118.1 Intron XP_016857607.1
XM_017002119.1 Intron XP_016857608.1
XM_017002120.1 Intron XP_016857609.1
XM_017002121.1 Intron XP_016857610.1
XM_017002122.1 Intron XP_016857611.1

View Full Product Details