Product Details

SNP ID
rs33912216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1793739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGGCTGAGGTGTCCGAGGCGCC[A/G]CCGCAGTCCTGGGCATTCTCCTGCG
Phenotype
MIM: 607946 MIM: 608171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADIPOR2 PubMed Links

Gene Details

Gene
ADIPOR2
Gene Name
adiponectin receptor 2
There are no transcripts associated with this gene.

Gene
CACNA2D4
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172364.4 3561 Silent Mutation GGC,GGT G1110G NP_758952.4
XM_011521041.2 3561 Intron XP_011519343.1

View Full Product Details